Learn More
Medchemexpress LLC HY-P1362 1mg Medchemexpress, Amyloid β Peptide (42-1)(human) CAS:317366-82-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP13621MG
Encompass_Preferred
Medchemexpress, HY-P1362 1mg Amyloid β Peptide (42-1)(human) CAS:317366-82-8 Amyloid β Peptide (42-1)(human) is the inactive form of Amyloid β Peptide (1-42). Amyloid β Peptide (42-1) is a 42-amino acid peptide which plays a key role in the pathogenesis of Alzheimer disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4191