Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMZ 30
GREENER_CHOICE

Catalog No. p-7250164
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-231-0 10 mg
69-235-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-231-0 Supplier Tocris Bioscience™ Supplier No. 6923/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective and irreversible protein phosphatase methylesterase-1 (PME-1) inhibitor

AMZ 30 is a potent, selective and irreversible inhibitor of protein phosphatase methylesterase-1 (PME-1; IC50 = 600 nM) with > 100-fold selectivity for PME-1 over other serine hydrolases in human cell lysates. AMZ 30 inactivates PME-1 and reduces the demethylated form of serine/threonine protein phosphatase 2A (PP2A) in HEK 293T cells. AMZ 30 perturbs leucine carboxyl methyltransferase-1 (LCMT1)-PME-1 methylation equilibrium leading to shortening of mitotic spindles and mitotic arrest in HeLa cells. It also shows antiproliferative properties in an in vitro model of endometrial carcinoma.

Specifications

Chemical Name or Material (2 E)-2-[(4-Fluorophenyl)sulfonyl]-3-[1-[(3-nitrophenyl)sulfonyl]-1 H-pyrrol-2-yl]-2-propenenitrile
CAS 1313613-09-0
Quantity 10 mg
Target Hydrolase Inhibitors
Molecular Formula C19H12FN3O6S2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.