Learn More
Cayman Chemical Anabasine (hydrochloride), 15251-47-5, 50 mg
SDP
Supplier:  Cayman Chemical 1820650
Encompass_Preferred
A natural, nicotine-related alkaloid that binds the same receptor as nicotine, the nicotinic acetylcholine α4β2 subtype (Ki = 38 nM) and increases nicotine-induced hyperactivity, sensitization, and intravenous self-administration
Catalog No. NC1468736
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.