Learn More
Selleck Chemical LLC Anagliptin
SDP
Supplier:  Selleck Chemical LLC S59095MG
Encompass_Preferred
Anagliptin is an orally active and highly selective dipeptidyl peptidase-4 (DPP-4) inhibitor and exerts anti-inflammatory effects on macrophages adipocytes and mouse livers by suppressing NF- B activation
Catalog No. 50-000-03892
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.