Learn More
Vernier™ Analog Extension Cable
SDPSCI_ED
Catalog No. S16147ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Analog Sensors
Catalog No. For Use With (Equipment)
S16147ND Analog Sensors
1 options
Catalog No. S16147ND Supplier Vernier™ Supplier No. EXTBTA
Add to Cart
Add to Cart
Includes
Analog Extension Cable
Most of our sensors have cables that are 1.5 m long. There are times, however, that you may wish to have a longer cable. The Analog Sensor Extension Cable can be used with any Vernier sensor that connects to an analog channel of LabPro, CBL 2, or CBL. Each cable is 2 meters in length.
  • For use with sensor that connect to an analog channel of LabPro, CBL 2, or CBL
  • Cable is 2 meters in length
Warranty and Services
5-year limited warranty
Specifications
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Analog Sensors
Length (Metric) 2 m
WARNING: Cancer - www.P65Warnings.ca.gov