Learn More
Cytiva Analyslide Petri Dish - Analyslide Petri dish (100/pkg)
Supplier:  Cytiva 7231
Encompass
For Transport, Storage, and Viewing Membrane Samples Rectangular base fits most microscope stages. Inner cover ring for secure closure.
Catalog No. NC1867589