Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Analyzer Cup

Catalog No. 07000801 Shop All Greiner Bio-One Products
Change view
Click to view available options
Volume (Metric):
1.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07-000-801 1.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-801 Supplier Greiner Bio-One Supplier No. 729101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Hitachi analytical system. Greiner Bio-One Analyzer Cup is used in wide variety of auto analyzer carousels.

  • Analyzer Cup for Hitachi
  • 17 x 38mm (0.6 x 1.4 in.)
TRUSTED_SUSTAINABILITY

Specifications

Color Crystal Clear
Product Type Analyzer Cup
Material Polystyrene
Volume (Metric) 1.7 mL
Sterility Non-sterile
For Use With (Application) Analytical systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.