Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vela Diagnostics Great Basin Analyzer - PA500- Refurbished

Catalog No. 23052001
Change view
Click to view available options
Certifications/Compliance:
FDA cleared platform
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
23-052-001 FDA cleared platform
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-052-001 Supplier Vela Diagnostics Supplier No. 370003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Automated bench-top analyzer offers both low-plex and multiplex, sample-to-result testing on a single system. The analyzer executes the assay, interprets the results, and provides electronic output, with results in under 2 hours. . The refurbished analyzer offers a reliable, cost-effective option.

  • An in vitro diagnostic device for use in detection of infectious disease targets.
  • The instrument includes electromechanical components that direct the flow of reagents in a disposable cartridge.
  • Resident software in the device determines test results which are automatically sent to the control platform—no interpretation of results is required.

Specifications

Humidity 20% to 80% RH non-condensing
Depth (English) 21.4 in.
Height (English) 17.2 in.
Certifications/Compliance FDA cleared platform
Content And Storage 20°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.