Learn More
ATCC Anaplasma marginale Theiler, strain South Idaho (S64-Id2AM) - Frozen rickettsiae reference strain (ATCC VR-1436)
Supplier:  ATCC VR1436
Encompass
ATCC VR-1436 is a reference strain of Anaplasma marginale (Theiler) derived from whole blood of a naturally infected Hereford cow in South Idaho (designation S64-Id2AM). Provided as frozen rickettsiae for veterinary hemoparasite research, the material is stored and shipped in the vapor phase of liquid nitrogen; infected blood remains viable for 24 hours at 4°C.
  • Reference strain suitable for veterinary hemoparasite research.
  • Strain designation: South Idaho (S64-Id2AM).
  • Isolated from whole blood of a naturally infected Hereford cow.
  • Provided as frozen rickettsiae and requires vapor-phase liquid nitrogen storage and shipment.
  • Viable in blood for up to 24 hours at 4°C.
  • Experimental challenge parameters: approximately 10^8 infected erythrocytes; 20-26 days by needle challenge (IV); 33-38 days by tick transmission.
Catalog No. NC2943487