Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Anastrozole, 98%

Catalog No. AC460970010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460970010 1 g
AC460972500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460970010 Supplier Thermo Scientific Chemicals Supplier No. 460970010
Add to Cart

CAS: 120511-73-1 | C17H19N5 | 293.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120511-73-1
Molecular Formula C17H19N5
Molecular Weight (g/mol) 293.37
InChI Key YBBLVLTVTVSKRW-UHFFFAOYSA-N
Synonym anastrozole, arimidex, anastrazole, anastrozol, 2,2'-5-1h-1,2,4-triazol-1-yl methyl-1,3-phenylene bis 2-methylpropanenitrile, anastrole, asiolex, zeneca zd 1033, unii-2z07myw1az, arimidex tn
PubChem CID 2187
ChEBI CHEBI:2704
IUPAC Name 2-[3-(2-cyanopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanenitrile
SMILES CC(C)(C#N)C1=CC(=CC(=C1)CN2C=NC=N2)C(C)(C)C#N

Specifications

Melting Point 81.0°C to 82.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 293.37
Percent Purity 98%
Chemical Name or Material Anastrozole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed. May damage fertility or the unborn child.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.