Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anastrozole, Tocris Bioscience™
SDP

Catalog No. 338850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-885-0 50 mg
33-881-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-885-0 Supplier R&D Systems Supplier No. 3388/50
This item is not returnable. View return policy

Potent aromatase (CYP19) inhibitor

Potent and highly selective aromatase (CYP19) inhibitor (IC50 = 15 nM) that has no discernible effect on adrenocorticoid hormone synthesis. Reduces plasma estrogen levels and exhibits antitumor activity in vivo. Orally active.

Chemical Identifiers

CAS 120511-73-1
Molecular Formula C17H19N5
Molecular Weight (g/mol) 293.374
InChI Key YBBLVLTVTVSKRW-UHFFFAOYSA-N
Synonym anastrozole, arimidex, anastrazole, anastrozol, 2,2'-5-1h-1,2,4-triazol-1-yl methyl-1,3-phenylene bis 2-methylpropanenitrile, anastrole, asiolex, zeneca zd 1033, unii-2z07myw1az, arimidex tn
PubChem CID 2187
ChEBI CHEBI:2704
IUPAC Name 2-[3-(2-cyanopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2-methylpropanenitrile
SMILES CC(C)(C#N)C1=CC(=CC(=C1)CN2C=NC=N2)C(C)(C)C#N

Specifications

Quantity 50 mg
Formula Weight 293.37
Percent Purity >99%
Chemical Name or Material Anastrozole
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.