Learn More
AdipoGen Ang-2h mAb blockAngy-1-4
SDP
Supplier:  AdipoGen AG27B0015C100
Encompass_Preferred
Recombinant Antibody. Recognizes human angiopoietin-2. Does not recognize mouse angiopoietin-2 and human angiopoietin-1. Species cross-reactivity Human. Clone Angy-1-4. Isotype Human IgG2lambda. Applications ELISA, FUNC Blocking. Host CHO cells. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Angiopoietin-1 Ang-1 and Angiopoietin-2 Ang-2 are closely related secreted ligands which bind with similar affinity to Tie-2. Tie-2 and angiopoietins have been shown to play critical roles in embryogenic angiogenesis and in maintaining the integrity of the adult vasculature. Ang-1 activates Tie-2 signaling on endothelial cells to promote chemotaxis, cell survival, cell sprouting, vessel growth and stabilization. Ang-2 has been identified as a secreted protein ligand of Tie-2 and has alternatively been reported to be an antagonist for Ang-1 induced Tie-2 signaling as well as an agonist for Tie-2 signaling, depending on the cell context.
Catalog No. 50-256-3152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.