Learn More
MyBioSource Mouse Angiotensin II (Ang II) ELISA Kit
SDP
Supplier:  MyBioSource MBS26843096STRIPWELLS
Encompass_Preferred
The MyBioSource Mouse Angiotensin II ELISA Kit quantitatively measures Angiotensin II in various mouse biological samples using a sandwich enzyme immunoassay. The 96-well plate coated with a specific antibody captures Angiotensin II, which is then detected with a biotinylated antibody and HRP-conjugated streptavidin. Color development, proportional to Angiotensin II levels, is measured at 450 nm post-TMB substrate addition and stop solution application.
Catalog No. NC2316021
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.