Learn More
AdipoGen anti-Angiopoietin-2, mAb (rec.) (blocking) (Angy-2-1) (100 µg)
SDP
Supplier:  AdipoGen AG27B0016C100
Encompass_Preferred
Recombinant Antibody. Recognizes human and mouse angiopoietin-2. Does not detect human angiopoietin-1. Species cross-reactivity: Human, Mouse. Clone: Angy-2-1. Isotype: Mouse IgG2blambda. Applications: ELISA, FUNC (Blocking). Host: CHO cells. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Ang-1 activates Tie-2 signaling on endothelial cells to promote chemotaxis, cell survival, cell sprouting, vessel growth and stabilization. Ang-2 has been identified as a secreted protein ligand of Tie-2 and has alternatively been reported to be an antagonist for Ang-1 induced Tie-2 signaling as well as an agonist for Tie-2 signaling, depending on the cell context. anti-Angiopoietin-2, mAb (rec.) (blocking) (Angy-2-1) is an antibody developed by antibody phage display technology using a human naive antibody gene library.
Catalog No. 50-205-3039
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.