Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Angiogenin Recombinant Rabbit Monoclonal Antibody (HL2215)
GREENER_CHOICE

Catalog No. FEMA550436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550436 Supplier Invitrogen™ Supplier No. MA550436
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Angiogenin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Angiogenin (ANG or ANG I) is important for the process of neovascularization and formation of new blood vessels. ANG is similar to pancreatic ribonuclease A and functions as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. It interacts with endothelial cell-surface actin and may cause changes in the cell cytoskeleton. ANG is thought to be involved in the development of solid tumors and its antagonists are capable of inhibiting tumor growth. Defects in ANG are the cause of susceptibility to amyotrophic lateral sclerosis type 9 (ALS9). Angiogenin is a genetic link between ALS and PD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Angiogenin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2215
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ANG
Gene Accession No. P03950
Gene Alias AI385586; ALS9; ANG; Ang1; Angiogenin; angiogenin ribonuclease 1; angiogenin, ribonuclease A family, member 1; angiogenin, ribonuclease, RNase A family, 5; angiogenin-1; epididymis luminal protein 168; HEL168; MGC22466; MGC71966; RAA1; Ribonuclease 5; ribonuclease A A1; ribonuclease A family member 5; RNase 5; RNASE4; RNASE5; Rnase5a
Gene Symbols ANG
Host Species Rabbit
Immunogen Recombinant fragmemt of human Angiogenin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283, 305843
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.