Learn More
Sino Biological Cynomolgus / Rhesus Angiopoietin-2 / ANG2 Protein (His Tag) 100ug
Supplier:  Sino Biological 90026C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus ANGPT2 (XP_005562586.1/XP_001097949.1) (Met1-Phe495) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus ANGPT2 sequences are identical.
Catalog No. NC1705575
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.