Learn More
MilliporeSigma™ Angiotensin Converting Enzyme, His·Tag™, Human, Recombinant, NSO Cells
Shop All MilliporeSigma ProductsDescription
- Recombinant, human ACE ectodomain (amino acids 30-1261) fused at the C-terminus to a His•Tag™ sequence and expressed in NSO cells
- ACE is a zinc metallopeptidase that cleaves the C-terminal dipeptide from angiotensin I to produce the potent vasopressor octapeptide angiotensin II
- Inactivates bradykinin by the sequential removal of two C-terminal dipeptides
- ACE is a type I membrane protein that exists in two isoforms, somatic and germinal
- By SDS-PAGE, the apparent molecular mass of the glycosylated protein appears larger than 120 kDa under either reducing or non-reducing condition
Specifications
Specifications
| Concentration | Concentration Label: Please refer to vial label for lot-specific concentration |
| Formulation | In 75mM NaCl, 12.5mM Tris, 500nM ZnCl₂, 40% glycerol, pH 7.5 |
| Purity | ≥95% by SDS-PAGE |
| Content And Storage | ≤-70°C |
| Form | Liquid |
| Quantity | 10 μg |
| Synonym | Peptidyl-Dipetidase A, ACE |
| Product Type | Angiotensin Converting Enzyme (ACE) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.