Learn More
CPC Scientific Glp-Gly-Leu-Pro-Pro-Gly-Pro-Pro-Ile-Pro-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ACEI003A
Encompass
Angiotensin I-converting enzyme inhibitor. Bradykinin Potentiator C.ONE-LETTER SEQUENCE: Glp-GLPPGPPIPPMOLECULAR FORMULA: C51H77N11O13MOLECULAR WEIGHT:1052.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: CardiovascularREFERENCES: H. Kato et al., Biochemistry, 10, 972 (1971)
Catalog No. 50-210-8578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.