Learn More
Angiotensin I Monoclonal Antibody (BGN/KA/4H), Invitrogen™
Description
This antibody does not react with angiotensin II, III, or angiotensinogen in ELISA. Mouse anti Human Angiotensin I, clone Ang E9, is specific for Angiotensin I and does not react with angiotensin II, III or angiotensinogen in ELISA.
Specifications
Specifications
| Antigen | Angiotensin I |
| Applications | ELISA, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | BGN/KA/4H |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide; pH 7.2 |
| Gene | AGT |
| Gene Accession No. | P01019 |
| Gene Alias | AGT; alpha-1 antiproteinase, antitrypsin; Ang I; Ang II; Ang III; Ang IV; Angiotensin 1-10; Angiotensin 1-4; Angiotensin 1-5; Angiotensin 1-7; Angiotensin 1-8; Angiotensin 1-9; Angiotensin 2-8; Angiotensin 3-8; Angiotensin I; angiotensin II; Angiotensin III; Angiotensin IV; Angiotensin-1; Angiotensin-2; Angiotensin-3; Angiotensin-4; Angiotensinogen; angiotensinogen (serpin peptidase inhibitor, clade A, member 8); ANHU; Des-Asp[1]-angiotensin II; FLJ92595; FLJ97926; pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; serpin A8; serpin peptidase inhibitor, clade A, member 8; SERPINA8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.