Learn More
Apexbio Technology LLC Angiotensin II 4474-91-3 250mg
SDP
Supplier:  Apexbio Technology LLC A1042250
Encompass_Preferred
Angiotensin II an endogenous octapeptide exerts potent vasoconstrictive effects through activation of angiotensin receptors on vascular smooth muscle cells This peptide stimulates contraction via binding G protein-coupled receptors triggering intracellular signaling cascades involving phospholipase C activation subsequent IP3-dependent calcium release and protein kinase C-mediated pathways Angiotensin II additionally promotes aldosterone secretion from adrenal cortical cells enhancing renal sodium and water reabsorption Experimentally angiotensin II is utilized extensively for investigating mechanisms of hypertension cardiovascular remodeling vascular smooth muscle cell hypertrophy and inflammatory responses in vascular injury models Reported IC50 values for receptor binding typically range between 1-10 nM depending on assay conditions
Catalog No. 50-000-63031
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.