Learn More
Enzo Life Sciences Angiotensin II ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900204
Encompass_Preferred
Highly sensitive ELISA, detecting as little as 4.6 pg/ml. The Angiotensin II EIA kit is a colorimetric, competitive immunoassay kit with results in 3.5 hours. Sensitivity: 4.6 pg/ml (range 3.9-10,000 pg/ml). Assay Time: 3.5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP
Catalog No. NC0583295
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.