Learn More
Medchemexpress LLC Angiotensin II human-13C6,15N TFA | 00-00-0 | MFCD00161484 | 98.1% | 1167.15 Da | C4613C6H72F3N1215NO14 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13948BS5MG
Encompass_Preferred
Angiotensin II human-13C6,15N TFA is a stable-isotope labeled form of the human bioactive peptide angiotensin II (sequence: DRVYIHPF), provided as the trifluoroacetic acid (TFA) salt. It is intended for use as an internal standard or tracer in quantitative mass spectrometry and bioanalytical workflows and is supplied in a 5 mg vial with high HPLC purity.
  • Isotopically labeled with 13C6 and 15N for accurate mass spectrometry quantitation.
  • Peptide sequence provided (DRVYIHPF) for unambiguous identification.
  • Supplied as the TFA salt to aid stability and solubility in common solvents.
  • High HPLC purity (98.11%) minimizes analytical interference.
  • Suitable for use as an internal standard, tracer, or reference in bioanalytical assays.
Catalog No. 50-002-22814