Learn More
Eurofins Discoverx Angiotensin II, Human, 5 mg. (92-1077)
Supplier: Eurofins Discoverx 921077
Angiotensin II is an endocrine, autocrine/paracrine, and intracrine hormone formed from the conversion of angiotensin I via angiotensin-converting enzyme (ACE). It increases blood pressure by vasoconstriction, sympathetic nervous stimulation, increased aldosterone biosynthesis, and Gαq protein stimulation in vascular smooth muscle. Also acts as a Na/H exchanger in the proximal tubules of the kidney, increasing blood sodium content and leading to increased blood volume, pressure and pH. Thus ACE inhibitors are considered anti-hypertensives. Provided as 5 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.