Learn More
CPC Scientific H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific RENI003A
Encompass
Renin substrate for human kidney.ONE-LETTER SEQUENCE: DRVYIHPFHLVIHMOLECULAR FORMULA: C79H116N22O17MOLECULAR WEIGHT:1645.93STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [82048-97-3], RESEARCH AREA: CardiovascularREFERENCES: D.A. Teksbury et al., BBRC, 99, 1311 (1981)
Catalog No. 50-211-0359
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.