Learn More
AdipoGen anti-ANGPTL3 (human), mAb (Kairos-37) (100 µg)
SDP
Supplier:  AdipoGen AG20A0039C100
Encompass_Preferred
Monoclonal Antibody. Recognizes the fibrinogen-like domain (FLD) of human ANGPTL3. Detects a band of ~68kDa and a cleaved band of ~35kDa by Western blot. Does not cross-react with other ANGPTL family proteins. Isotype: Mouse IgG1kappa. Clone: Kairos-37. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL3 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism.
Catalog No. 50-205-2833
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.