Learn More
AdipoGen ANGPTL4 h mAb Kairos-1
SDP
Supplier:  AdipoGen AG20A0038C050
Encompass_Preferred
Monoclonal Antibody. Recognizes the fibrinogen-like domain FLD of human ANGPTL4. Detects a band of ~62kDa and ~35kDa by Western blot. Does not cross-react with other ANGPTL family proteins. Isotype Mouse IgG1kappa. Clone Kairos-1. Applications ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL4 mainly expressed in endothelial cells hypoxia-induced. Regulates angiogenesis and modulates tumorgenesis and directly regulates lipid, glucose, and energy metabolism. Inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage. ANGPTL4 is a protein consisting of an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain FLD. Both domains have distinct biological functions. The coiled-coil domain is responsible for the inhibitory effects on lipoprotein lipase LPL converting the active form of LPL into an inactive form, and the FLD domain mediates its antiangiogenic functions.
Catalog No. 50-256-3391
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.