Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANGPTL4 Recombinant Superclonal™ Antibody (1HCLC)

Catalog No. PI710186 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI710186 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI710186 Supplier Invitrogen™ Supplier No. 710186
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Superclonal Antibody

Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL4
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 1HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene ANGPTL4
Gene Accession No. Q9BY76, Q9Z1P8
Gene Alias 425O18-1; angiopoietin like 4; angiopoietin-like 4; angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; ANGPTL4; ANGPTL4 C-terminal chain; ANGPTL4 N-terminal chain; ARP4; Bk89; Farp; Fasting-i; Fasting-induced adipose factor; FIAF; fibrinogen/angiopoietin-related protein; HARP; hepatic angiopoietin-related protein; hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Ng27; NL2; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PGAR; Pgarg; Pp1158; PPARG angiopoietin related protein; PSEC0166; Secreted protein Bk89; TGQTL; UNQ171; UNQ171/PRO197
Gene Symbols ANGPTL4
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 258-273 of human Angiopoietin-like 4.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51129, 57875
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.