Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANGPTL4 Recombinant Rabbit Monoclonal Antibody (1H12L19)

Catalog No. 701155
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-115-5 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-115-5 Supplier Invitrogen™ Supplier No. 701155
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1H12L19
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene ANGPTL4
Gene Accession No. Q9BY76, Q9Z1P8
Gene Alias 425O18-1; angiopoietin like 4; angiopoietin-like 4; angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; ANGPTL4; ANGPTL4 C-terminal chain; ANGPTL4 N-terminal chain; ARP4; Bk89; Farp; Fasting-i; Fasting-induced adipose factor; FIAF; fibrinogen/angiopoietin-related protein; HARP; hepatic angiopoietin-related protein; hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Ng27; NL2; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PGAR; Pgarg; Pp1158; PPARG angiopoietin related protein; PSEC0166; Secreted protein Bk89; TGQTL; UNQ171; UNQ171/PRO197
Gene Symbols ANGPTL4
Host Species Rabbit
Immunogen Human Angptl4.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51129, 57875
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.