Learn More
Abcam Anhydrotetracycline Hydrochloride, Tetracycline Repressor Effector, >98% Purity, 500 mg
Description
- Molecular Weight: 462.9 Da
- Purity: >98%
- Form: Solid
- Function: Acts as a tetracycline repressor and reverse tetracycline repressor effector.
- Applications: Used as a tetracycline-dependent gene expression effector in bacterial expression systems.
- Antibiotic Activity: Exhibits no antibiotic activity and binds poorly to the 30S ribosomal subunit.
- Derivative: Tetracycline (ab141223, ab146579) derivative.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Tetracycline Repressor |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 98% anhydrotetracycline hydrochloride |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 98% Anhydrotetracycline hydrochloride |
| Quantity | 500 mg |
| For Use With (Application) | Tetracycline-dependent gene expression |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.