Learn More
Abcam Anhydrotetracycline Hydrochloride, Tetracycline Repressor Effector, Solid, 2500 mg
Description
- Molecular Weight: 462.9 Da
- Purity: >98%
- Physical Form: Solid
- Function: Tetracycline repressor and reverse tetracycline repressor effector; tetracycline-dependent gene expression effector in bacterial expression systems
- Properties: Tetracycline derivative; binds poorly to the 30S ribosomal subunit; exhibits no antibiotic activity
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Tetracycline Repressor |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Buffer | 98% anhydrotetracycline hydrochloride |
| Formulation | Constituents: 98% Anhydrotetracycline hydrochloride |
| Quantity | 2,500 mg |
| For Use With (Application) | Tetracycline-dependent gene expression effector |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.