Learn More
TARGETMOL CHEMICALS INC ANI9 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T266305MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ani9 is a high selective blocker of Anoctamin1 (ANO1)/transmembrane protein 16A (TMEM16A) with an IC50 of 77 nM and can be used in studies about ANO1 and the treatment of ancer hypertension pain diarrhea and asthma. purity: 98%
Catalog No. 50-258-0682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.