Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline-2-sulfonic Acid, 95%

Catalog No. AC400641000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400641000 100 g Glass bottle
AC400640250 25 g Glass bottle
AC400645000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC400641000 Supplier Thermo Scientific Chemicals Supplier No. 400641000
  • $128.80
    $97.65 / Each of 1

    Save $31.15

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

CAS: 88-21-1 | C6H7NO3S | 173.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-21-1
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.19
MDL Number MFCD00007705
InChI Key ZMCHBSMFKQYNKA-UHFFFAOYSA-N
Synonym orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
PubChem CID 6926
ChEBI CHEBI:1015
IUPAC Name 2-aminobenzenesulfonic acid
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)O

Specifications

Melting Point >300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2NC6H4SO3H
Quantity 100 g
Beilstein 14, 681
Merck Index 15, 6978
Solubility Information Solubility in water: 3100mg/l (23°C)
Formula Weight 173.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Aniline-2-sulfonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 201-810-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.