Learn More
Johannes Lieder Animal Cell Set
SCI_ED
Catalog No. S5000 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Quantity:
12
Catalog No. Quantity
S5000 12
1 options
This item is not returnable. View return policy
Catalog No. S5000 Supplier Johannes Lieder Supplier No. 5000
This item is not returnable. View return policy
Twelve slides show a variety of tissues.
Set contains 12 slides of animal cytology with depictured accompanying brochure. Epithelium, muscle, bone, nerve, internal organs, sex organs and chromosomes are all represented in this slide set. It shows a fine cross section of tissue types useful for classroom microscope study.
Includes:
  • Squamous epithelium, isolated cells from human mouth. Nuclei and cytoplasm are shown
  • Striated muscle l.s. showing nuclei, striations, myofibrils
  • Compact bone and hyaline cartilage t.s., two sections on one slide for comparison
  • Nerve fibres isolated, fixed and stained by osmic acid to show myeline sheaths and Ranvier's nodes
  • Liver of Salamandra t.s., showing simple animal cells with cellular membranes, nuclei, and cytoplasm
  • Kidney of mouse, t.s. vital stained with trypanblue to demonstrate the storage of epithelial cells
  • Ovary of cat, t.s. showing primary, secondary, and Graafian follicles
  • Testis of frog, t.s. showing spermatogenesis. Spermatogonia, spermatocytes, spermatids, and mature spermatozoa
  • Salamandra larva, t.s. of skin and other organs selected to show cell division (mitosis) in various stages
  • Uteri of Ascaris megalocephala, t.s. iron hematoxyline stained to show details of meiosis with chromosomes and nuclear spindles
  • Salivary gland of Chironomus larva. Giant chromosomes showing large chromomeres. Stained for DNA after Feulgen
  • Ova from Psammechinus (sea urchin). Unfertilized ova, fertilized ova, early cleavage stages
Specifications
Type The Animal Cell
Quantity 12