Learn More
Vector Laboratories Animal-Free Blocker (5x) 250 ml
SDP
Supplier:  Vector Laboratories SP5030250
Encompass_Preferred
Animal-Free Blocker is a plant-derived blocking agent and diluent for immunohistochemistry, nucleic acid and protein blotting, and other applications. This reagent contains no animal-derived protein and can be used as an alternative to sera, BSA, casein, or non-fat dry milk. It is supplied as a 5x concentrate.
Catalog No. NC9592609
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.