Learn More
Stoelting™ Animal Markers
SDP
Catalog No. 10001076 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
Fine tip
Standard tip
Model:
Black
Blue
Green
Orange
Red
Yellow
Includes:
Pack of 2
Pack of 3
Single Packaging
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Model Includes
10-001-076 Standard tip Yellow Single Packaging
10-000-646 Fine tip Black Pack of 3
10-001-084 Fine tip Black Single Packaging
10-000-645 Standard tip Black Pack of 2
10-001-083 Standard tip Black Single Packaging
10-000-640 Fine tip Blue Pack of 3
10-001-078 Fine tip Blue Single Packaging
10-000-635 Standard tip Blue Pack of 2
10-001-073 Standard tip Blue Single Packaging
10-000-641 Fine tip Green Pack of 3
10-001-079 Fine tip Green Single Packaging
10-000-636 Standard tip Green Pack of 2
10-001-074 Standard tip Green Single Packaging
10-000-644 Fine tip Orange Pack of 3
10-001-082 Fine tip Orange Single Packaging
10-000-639 Standard tip Orange Pack of 2
10-001-077 Standard tip Orange Single Packaging
10-000-642 Fine tip Red Pack of 3
10-001-080 Fine tip Red Single Packaging
10-000-637 Standard tip Red Pack of 2
10-001-075 Standard tip Red Single Packaging
10-000-643 Fine tip Yellow Pack of 3
10-001-081 Fine tip Yellow Single Packaging
10-000-638 Standard tip Yellow Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 10-001-076 Supplier Stoelting™ Supplier No. 504431
Add to Cart
Add to Cart
Animal Markers allow for easy, long-lasting identification of laboratory animals.
  • Nontoxic and devoid of any hazardous materials
  • A single marker can be used to mark 300 to 500 rats (depending upon animal size and/or applied area)
  • Once applied, colors remain for 6 to 12 weeks
  • Broad and fine tip markers available
Specifications
For Use With (Application) Neuroscience
Size Standard tip
Animal Rodent
Model Yellow
Includes Single Packaging