Learn More
- Home
- Products
- Animal Care and Research
- Animal Surgical Devices
- Animal Surgical Equipment
- Stoelting™ Animal Markers
Stoelting™ Animal Markers
Catalog No. 10001081
Shop All Stoelting Co Products
Change view
Click to view available options
Size:
Fine tip
Standard tip
Model:
Black
Blue
Green
Orange
Red
Yellow
Includes:
Pack of 2
Pack of 3
Single Packaging
24 product options available for selection
| Catalog No. | Size | Model | Includes |
|---|---|---|---|
| 10-001-081 | Fine tip | Yellow | Single Packaging |
| 10-000-646 | Fine tip | Black | Pack of 3 |
| 10-001-084 | Fine tip | Black | Single Packaging |
| 10-000-645 | Standard tip | Black | Pack of 2 |
| 10-001-083 | Standard tip | Black | Single Packaging |
| 10-000-640 | Fine tip | Blue | Pack of 3 |
| 10-001-078 | Fine tip | Blue | Single Packaging |
| 10-000-635 | Standard tip | Blue | Pack of 2 |
| 10-001-073 | Standard tip | Blue | Single Packaging |
| 10-000-641 | Fine tip | Green | Pack of 3 |
| 10-001-079 | Fine tip | Green | Single Packaging |
| 10-000-636 | Standard tip | Green | Pack of 2 |
| 10-001-074 | Standard tip | Green | Single Packaging |
| 10-000-644 | Fine tip | Orange | Pack of 3 |
| 10-001-082 | Fine tip | Orange | Single Packaging |
| 10-000-639 | Standard tip | Orange | Pack of 2 |
| 10-001-077 | Standard tip | Orange | Single Packaging |
| 10-000-642 | Fine tip | Red | Pack of 3 |
| 10-001-080 | Fine tip | Red | Single Packaging |
| 10-000-637 | Standard tip | Red | Pack of 2 |
| 10-001-075 | Standard tip | Red | Single Packaging |
| 10-000-643 | Fine tip | Yellow | Pack of 3 |
| 10-000-638 | Standard tip | Yellow | Pack of 2 |
| 10-001-076 | Standard tip | Yellow | Single Packaging |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
Description
- Nontoxic and devoid of any hazardous materials
- A single marker can be used to mark 300 to 500 rats (depending upon animal size and/or applied area)
- Once applied, color remains on fur for 6 to 12 weeks; does not work on bare skin
- Broad and fine tip markers available
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Size | Fine tip |
| Animal | Rodent |
| Model | Yellow |
| Includes | Single Packaging |