Learn More
Walter Products™ Animal Meiosis Model
SCI_ED
Catalog No. S139168 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
23.6 x 15.75 x 2.4 in.
Catalog No. Dimensions (L x W x H)
S139168 23.6 x 15.75 x 2.4 in.
1 options
This item is not returnable. View return policy
Catalog No. S139168 Supplier Walter Products™ Supplier No. B10509
Add to Cart
Add to Cart
This item is not returnable. View return policy
This animal meiosis model provides a three-dimensional view for study of the 10-phase cell division process of mammal cell.
  • Shows Interphase (stage of G1-phase), Prophase I (leptotene), Prophase I (zygotene and pachytene), Prophase I (diplotene), Prophase I (diakinesis), Metaphase I, Anaphase I, [Telophase I, cytokinesis I, interkinesis, prophase II and metaphase II], Anaphase II, Telophase II and Cytokinesis II
  • Model is enlarged 10,000 times for accurate life size demonstration
  • Each of the 10 pieces is equiped with Velcro to attach to the base and the base can be fastened to a wall for versatile display and a better teaching experience
  • Animal meiosis model is colored to microscopy and biology standards for accurate identification and equipped with polyethylene stand for fast and easy attachment
  • Detailed open cell organelles for easy teaching identification
Specifications
Dimensions (L x W x H) 23.6 x 15.75 x 2.4 in.
Includes 10 Pieces
Material PVC
Product Type Anatomical Model