Learn More
Walter Products™ Animal Mitosis Model
SCI_ED
Catalog No. S139169 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
23.6 x 15.75 x 2.4 in.
Catalog No. Dimensions (L x W x H)
S139169 23.6 x 15.75 x 2.4 in.
1 options
This item is not returnable. View return policy
Catalog No. S139169 Supplier Walter Products™ Supplier No. B10510
Add to Cart
Add to Cart
This item is not returnable. View return policy
This animal mitosis model provides a three-dimensional view for the study of the 9-phase cell division process of a mammal cell.
  • Shows Interphase, Prophase, Early Prometaphase, Late Prometaphase, Metaphase, Early Anaphase, Late Anaphase, Telophase, Cytokinesis
  • Model is enlarged 10,000 times for accurate life size demonstration
  • Each of the 9 pieces is equiped with Velcro to attach to the base and the base can be fastened to a wall for versatile display and a better teaching experience
  • Animal mitosis model is colored to microscopy and biology standards for accurate identification and equipped with polyethylene stand for fast and easy attachment
  • Detailed open cell organelles for easy teaching identification
Specifications
Dimensions (L x W x H) 23.6 x 15.75 x 2.4 in.
Includes 9 Pieces
Material PVC
Product Type Anatomical Model