Learn More
STEMCELL Technologies TeSR™-AOF
SDP
Supplier:  STEMCELL Technologies 1000401
Encompass
Reduce risk and obtain high-quality cells in your human pluripotent stem cell-derived (hPSC) cell therapy development with TeSR™-AOF, manufactured under relevant cGMPs. With no materials of animal or human origin to the secondary level of manufacturing, enjoy more straightforward traceability and enhanced viral safety compared to media that are only animal origin-free to the primary level of manufacturing.To enhance cell quality attributes, particularly during restricted feeds, critical medium components have been stabilized, including fibroblast growth factor 2 (FGF2 aka bFGF).TeSR™-AOF is compatible with a variety of culture matrices, including Corning Matrigel hESC-Qualified Matrix, Vitronectin XF™ (Catalog #07180), and CellAdhere™ Laminin-521 (Catalog #77003).For quality information visit www.stemcell.com/compliance. Request a Letter of Authorization for the FDA Master File www.stemcell.com/loa-request-dmf.
Catalog No. 50-206-9320