Learn More
Caron Products And Services Animal Rearing Chamber, 25 cu. ft., LED Lights, Temp, No RH
Supplier:  Caron Products And Services 7350251
Encompass_Preferred
Carons 25 cu. ft. (708 liters) Animal Rearing Chamber. Standard features include touchscreen controls, temperature range from 10C to 40C, LED lighting, and light tight door cover. These chambers allow you to simulate specialized environments for your research needs. 115V, 60 Hz.
Catalog No. NC2676814
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.