Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anisole, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019073 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9073 10 mL
11-101-9907 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9073 Supplier MilliporeSigma Supelco Supplier No. 9610910MLF
Add to Cart
Add to Cart

Anisole is a colorless liquid , which can be obtained upon heating anisic acid with an excess of barium hydroxide.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-66-3
Ignition Point 887°F
Vapor Pressure 10 mmHg (42.2°C)
Vapor Density 3.7 (vs. air)
Density 0.995 g/mL (at 25°C)
Flash Point 43°C
Physical Form Neat
Linear Formula CH3 OC6H5
MDL Number MFCD00008354
Melting Point -37°C
Boiling Point 154°C
Synonym Methoxybenzene; Methyl phenyl ether
Type Analytical Standard
Quantity 10 mL
Formula Weight 108.14
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.