Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anisole, 99%, pure

Catalog No. AC153920025
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153920025 2.5 L Glass bottle
AC153920050 5 mL Glass bottle
AC153922500 250 mL Glass bottle
AC153920010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC153920025 Supplier Thermo Scientific Chemicals Supplier No. 153920025
  • $246.50
    $209.65 / Each of 1

    Save $36.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-66-3 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-66-3
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
Synonym methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
PubChem CID 7519
ChEBI CHEBI:16579
IUPAC Name anisole
SMILES COC1=CC=CC=C1

Specifications

Melting Point -37.0°C
Color Colorless
Density 0.9900g/mL
Boiling Point 154.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5160 to 1.5190
Linear Formula C6H5OCH3
Quantity 2.5 L
Beilstein 06,138
Fieser 01,41
Merck Index 15,662
Specific Gravity 0.99
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
Formula Weight 108.14
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Anisole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ven
EINECSNumber 202-876-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.