Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anisomycin, Protein Synthesis Inhibitor, Solid, 50 mg

Catalog No. 502707549
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7549 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7549 Supplier Abcam Supplier No. AB12049550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent translational inhibitor of protein synthesis that activates stress-activated protein kinases and JNK MAP kinase cascades.

  • Molecular Weight: 265.3 Da
  • Purity: >98%
  • Form: Solid
  • Function: Translational inhibitor of protein synthesis; highly potent activator of kinase cascades, including p38 and JNK MAP kinases
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Protein
Shipping Condition Ambient
Content And Storage Store at 4°C.
Form Solid
Formulation 100% Anisomycin
Quantity 50 mg
For Use With (Application) Protein synthesis inhibition, Kinase cascade activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.