Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anisomycin, Protein Synthesis Inhibitor, Solid, 98% Purity, 10 mg

Catalog No. 502707550
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7550 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7550 Supplier Abcam Supplier No. AB12049510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent translational inhibitor of protein synthesis and activator of kinase cascades, including p38 and JNK MAP kinases.

  • Molecular Weight: 265.3 Da
  • Purity: >98%
  • Description: Translational inhibitor of protein synthesis. Highly potent activator of kinase cascades. Potently activates stress-activated protein kinases, p38, JNK MAP and other kinases.
  • Form: Solid
  • Storage: Store at +4°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Protein, p38, JNK
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Formulation 100% Anisomycin
Quantity 10 mg
For Use With (Application) Protein synthesis inhibition, Kinase cascade activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.