Learn More
ANKIB1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301367A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Might act as an E3 ubiquitin-protein ligase, or as part of E3 complex, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes and then transfers it to substrates.Specifications
| ANKIB1 | |
| Polyclonal | |
| Unconjugated | |
| ANKIB1 | |
| ankyrin repeat and IBR domain-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54467 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9P2G1 | |
| ANKIB1 | |
| Between 925 and 975 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.