Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKLE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572735
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572735 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572735 Supplier Invitrogen™ Supplier No. PA572735
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

ANKLE1 Polyclonal Antibody for Western Blot, ELISA

Endonuclease that probably plays a role in the DNA damage response and DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKLE1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ANKLE1
Gene Accession No. A8VU90, Q8NAG6
Gene Alias 8430438L13Rik; Ankle1; Ankrd41; ankyrin repeat and LEM domain containing 1; ankyrin repeat and LEM domain-containing protein 1; ankyrin repeat domain 41; ankyrin repeat domain-containing protein 41; C430003I23; LEM domain containing 6; Lem3; LEMD6; LEM-domain containing 3; LEM-domain containing protein 3; RGD1308184
Gene Symbols ANKLE1
Host Species Chicken
Immunogen ANKLE1 antibody for a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is within the last 50 amino acids of ANKLE1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126549, 234396, 361122
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.