Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKLE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109389 Supplier Invitrogen™ Supplier No. PA5109389
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CRYNVMHVAA KENQASICQL TLDVLENPDF MRLMYPDDDE AMLQKRIRYV VDLYLNTPDK MGYDTPLHFA CKFGNADVVN VLSSHHLIVK NSRNKYDKTP E.

This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKLE2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ANKLE2
Gene Accession No. Q6P1H6, Q7TP65, Q86XL3
Gene Alias 1110001J12Rik; Ab2-034; AI661024; Ankle2; ankyrin repeat and LEM domain containing 2; ankyrin repeat and LEM domain-containing protein 2; D5Ertd585e; Kiaa0692; LEM domain containing 7; LEM domain-containing protein 4; LEM4; LEMD7; liver regeneration-related protein LRRG057; MCPH16; RGD1310191
Gene Symbols ANKLE2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-450 of human ANKLE2 (NP_055929.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23141, 360829, 71782
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.