Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD11 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501736124 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6124 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6124 Supplier Proteintech Group Inc Supplier No. 553641AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody is specific to ANKRD11.

ANKRD11 is a member of a novel family of ankyrin repeats containing cofactors (ANCOs) that interact with p160 coactivators to inhibit ligand-dependent transactivation. ANKRD11 encodes a large nuclear protein with five ankyrin repeats, and parts of its sequences have been reported as nasopharyngeal carcinoma susceptibility protein and medulloblastoma antigen. This gene also colocalizes and interacts with histone deacetylases.

Specifications

Antigen ANKRD11
Applications Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANKRD11
Gene Accession No. Q6UB99
Gene Alias ANCO 1, ANCO1, ANKRD11, ankyrin repeat domain 11, LZ16, T13
Gene Symbols ANKRD11
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29123
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.