Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD13C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569537 Supplier Invitrogen™ Supplier No. PA569537
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD13C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRD13C
Gene Accession No. Q3UX43
Gene Alias AI505652; Ankrd13c; ankyrin repeat domain 13c; ankyrin repeat domain-containing protein 13C; AU022220; dJ677H15.3; fc57c01; RP4-677H15.5; similar to hypothetical protein DKFZp566D1346; wu:fc57c01; wu:fq56c05; zgc:56077
Gene Symbols ANKRD13C
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse ANKRD13C.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 433667
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.