Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540110 Supplier Invitrogen™ Supplier No. PA540110
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: human muscle, mouse muscle, rat muscle tissue. Predicted reactivity: Mouse (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Ankyrin repeat domain 33 (ANKRD33), also named as C12orf7, belongs to the ankyrin repeat family and is highly expressed in retina. ANKRD33 exists as two isoforms. The isoform 1 molecular weight is 29 kDa and isoform 2 is 49 kDa. The function of ANKRD33 is still non-known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD33
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% Thimerosal; pH 7
Gene ANKRD33
Gene Accession No. Q7Z3H0, Q8BXP5
Gene Alias A930021G21Rik; Ankrd33; ankyrin repeat domain 33; ankyrin repeat domain-containing protein 33; C12orf7; LOC683410; PANKY; Panky-a; Photoreceptor ankyrin repeat protein; photorecptor ankyrin repeat protein A; similar to ankyrin repeat domain 33
Gene Symbols ANKRD33
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ANKRD33. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208258, 341405, 683410
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.